Anti-NCOA3 Mouse Monoclonal Antibody [clone: AX 15]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB12074-50EA 4140 DKK
ABCAAB12074-50
Anti-NCOA3 Mouse Monoclonal Antibody [clone: AX 15]
Type: Primary
Antigen: NCOA3 (nuclear receptor coactivator 3)
Clonality: monoclonal
Clone: AX 15
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR