Anti-AKT1 Mouse Monoclonal Antibody [clone: 104A282]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11901-50EA 4410 DKK
ABCAAB11901-50
Anti-AKT1 Mouse Monoclonal Antibody [clone: 104A282]
Type: Primary
Antigen: AKT1 (v-akt murine thymoma viral oncogene homolog 1)
Clonality: monoclonal
Clone: 104A282
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR