Anti-C3 / C3a Mouse Monoclonal Antibody [clone: 2898]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11874-50EA 5670 DKK
ABCAAB11874-50
Anti-C3 / C3a Mouse Monoclonal Antibody [clone: 2898]
Antistoffer
Type: Primary
Antigen: C3 / C3a
Clonality: monoclonal
Clone: 2898
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR