Anti-E2F4 Mouse Monoclonal Antibody [clone: WUF3]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11845-100EA 4650 DKK
ABCAAB11845-100
Anti-E2F4 Mouse Monoclonal Antibody [clone: WUF3]
Type: Primary
Antigen: E2F4 (E2F transcription factor 4, p107/p130-binding)
Clonality: monoclonal
Clone: WUF3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR