Anti-COIL Mouse Monoclonal Antibody [clone: Pdelta]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11822-50EA 5810 DKK
ABCAAB11822-50
Anti-COIL Mouse Monoclonal Antibody [clone: Pdelta]
Antistoffer
Type: Primary
Antigen: COIL (coilin)
Clonality: monoclonal
Clone: Pdelta
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR