Anti-URO7 Mouse Monoclonal Antibody [clone: S22]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11772-500EA 4250 DKK
ABCAAB11772-500
Anti-URO7 Mouse Monoclonal Antibody [clone: S22]
Type: Primary
Antigen: URO7
Clonality: monoclonal
Clone: S22
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR