Anti-MPO Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 2C7]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11729-50EA 5630 DKK
ABCAAB11729-50
Anti-MPO Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 2C7]
Antistoffer
Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: monoclonal
Clone: 2C7
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR