Anti-IgM Mouse Monoclonal Antibody [clone: MARM-4] (HRP (Horseradish Peroxidase))
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB11678-250EA 2011.96 DKK
ABCAAB11678-250
Anti-IgM Mouse Monoclonal Antibody [clone: MARM-4] (HRP (Horseradish Peroxidase))
Antistoffer
Type: Secondary
Antigen: IgM (Rat) (rat IgM)
Clonality: monoclonal
Clone: MARM-4
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu