Anti-MMP12 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11612-25EA 4140 DKK
ABCAAB11612-25
Anti-MMP12 Rabbit Polyclonal Antibody
Type: Primary
Antigen: MMP12 (matrix metallopeptidase 12 (macrophage elastase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR