Anti-VTN Mouse Monoclonal Antibody [clone: VIT-2]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11591-50EA 4510 DKK
ABCAAB11591-50
Anti-VTN Mouse Monoclonal Antibody [clone: VIT-2]
Type: Primary
Antigen: VTN (vitronectin)
Clonality: monoclonal
Clone: VIT-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR