Anti-IgG1 Rat Monoclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11585-250EA 2280 DKK
ABCAAB11585-250
Anti-IgG1 Rat Monoclonal Antibody
Antistoffer
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR