Anti-IgE Rat Monoclonal Antibody [clone: LO-ME-3] (Biotin)

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11580-250EA 2650 DKK
ABCAAB11580-250
Anti-IgE Rat Monoclonal Antibody [clone: LO-ME-3] (Biotin)
Antistoffer
Type: Secondary
Antigen: IgE (Mouse) (mouse IgE)
Clonality: monoclonal
Clone: LO-ME-3
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR