Anti-Laminin 2 alpha Rat Monoclonal Antibody [clone: 4H8-2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11576-25EA 5790 DKK
ABCAAB11576-25
Anti-Laminin 2 alpha Rat Monoclonal Antibody [clone: 4H8-2]
Antistoffer
Type: Primary
Antigen: Laminin 2 alpha
Clonality: monoclonal
Clone: 4H8-2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR