Anti-PSMD7 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11436-50EA 5810 DKK
ABCAAB11436-50
Anti-PSMD7 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PSMD7 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR