Anti-ITGAM Mouse Monoclonal Antibody FITC (Fluorescein) [clone: ICRF44]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11356-50EA 1810 DKK
ABCAAB11356-50
Anti-ITGAM Mouse Monoclonal Antibody FITC (Fluorescein) [clone: ICRF44]
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: monoclonal
Clone: ICRF44
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR