Anti-Mucin 5AC Mouse Monoclonal Antibody [clone: 45M1]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11335-200EA 0 DKK
ABCAAB11335-200
Anti-Mucin 5AC Mouse Monoclonal Antibody [clone: 45M1]
Type: Primary
Antigen: Mucin 5AC
Clonality: monoclonal
Clone: 45M1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR