Anti-alpha Adaptin Mouse Monoclonal Antibody [clone: 100/2]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11326-100EA 4360 DKK
ABCAAB11326-100
Anti-alpha Adaptin Mouse Monoclonal Antibody [clone: 100/2]
Type: Primary
Antigen: alpha Adaptin
Clonality: monoclonal
Clone: 100/2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR