Anti-TUBB Mouse Monoclonal Antibody [clone: TUB 2.1]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11308-200EA 4360 DKK
ABCAAB11308-200
Anti-TUBB Mouse Monoclonal Antibody [clone: TUB 2.1]
Type: Primary
Antigen: TUBB (tubulin, beta class I)
Clonality: monoclonal
Clone: TUB 2.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR