Anti-hCG alpha 4 Mouse Monoclonal Antibody [clone: INN-hFSH-132]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11232-250EA 3880.22 DKK
ABCAAB11232-250
Anti-hCG alpha 4 Mouse Monoclonal Antibody [clone: INN-hFSH-132]
Antistoffer
Type: Primary
Antigen: hCG alpha 4
Clonality: monoclonal
Clone: INN-hFSH-132
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR