Anti-MBP Mouse Monoclonal Antibody [clone: 1 (129-138)]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11160-1EA 4650 DKK
ABCAAB11160-1
Anti-MBP Mouse Monoclonal Antibody [clone: 1 (129-138)]
Type: Primary
Antigen: MBP (myelin basic protein)
Clonality: monoclonal
Clone: Clone 1 (129-138)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR