Anti-MYO5A Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11094-100EA 4650 DKK
ABCAAB11094-100
Anti-MYO5A Rabbit Polyclonal Antibody
Type: Primary
Antigen: MYO5A (myosin VA (heavy chain 12, myoxin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR