Anti-COMP Rat Monoclonal Antibody [clone: MA37C94 (HC484D1)]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11056-1EA 4360 DKK
ABCAAB11056-1
Anti-COMP Rat Monoclonal Antibody [clone: MA37C94 (HC484D1)]
Type: Primary
Antigen: COMP (cartilage oligomeric matrix protein)
Clonality: monoclonal
Clone: MA37C94 (HC484D1)
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR