Anti-RAB11A Mouse Monoclonal Antibody [clone: 7C10]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11034-50EA 5630 DKK
ABCAAB11034-50
Anti-RAB11A Mouse Monoclonal Antibody [clone: 7C10]
Antistoffer
Type: Primary
Antigen: RAB11A (RAB11A, member RAS oncogene family)
Clonality: monoclonal
Clone: 7C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR