Anti-SLIT3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11018-100EA 5760 DKK
ABCAAB11018-100
Anti-SLIT3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLIT3 (slit homolog 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR