Anti-ACTN Mouse Monoclonal Antibody [clone: BM 75.2]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB11008-100EA 4510 DKK
ABCAAB11008-100
Anti-ACTN Mouse Monoclonal Antibody [clone: BM 75.2]
Type: Primary
Antigen: ACTN (actinin, alpha)
Clonality: monoclonal
Clone: BM 75.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Chicken, Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR