Anti-KDR Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10972-25EA 5620 DKK
ABCAAB10972-25
Anti-KDR Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KDR (kinase insert domain receptor (a type III receptor tyrosine kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Dog, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR