Anti-ETS1 Mouse Monoclonal Antibody [clone: 1G11]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10936-250EA 4460 DKK
ABCAAB10936-250
Anti-ETS1 Mouse Monoclonal Antibody [clone: 1G11]
Type: Primary
Antigen: ETS1 (v-ets avian erythroblastosis virus E26 oncogene homolog 1)
Clonality: monoclonal
Clone: 1G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR