Anti-IRF6 Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10925-100EA 5330 DKK
ABCAAB10925-100
Anti-IRF6 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IRF6 (interferon regulatory factor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR