Anti-NOV Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10888-100EA 4480 DKK
ABCAAB10888-100
Anti-NOV Rabbit Polyclonal Antibody
Type: Primary
Antigen: NOV (nephroblastoma overexpressed)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR