Anti-BMPR2 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10862-50EA 4360 DKK
ABCAAB10862-50
Anti-BMPR2 Goat Polyclonal Antibody
Type: Primary
Antigen: BMPR2 (bone morphogenetic protein receptor, type II (serine/threonine kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR