Anti-TGF beta 2 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10850-50EA 4360 DKK
ABCAAB10850-50
Anti-TGF beta 2 Goat Polyclonal Antibody
Type: Primary
Antigen: TGF beta 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR