Anti-IL9R Mouse Monoclonal Antibody [clone: 33423]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10773-100EA 4250 DKK
ABCAAB10773-100
Anti-IL9R Mouse Monoclonal Antibody [clone: 33423]
Type: Primary
Antigen: IL9R (interleukin 9 receptor)
Clonality: monoclonal
Clone: 33423
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR