Anti-POU1F1 Mouse Monoclonal Antibody [clone: 2C11]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10623-500EA 4172.96 DKK
ABCAAB10623-500
Anti-POU1F1 Mouse Monoclonal Antibody [clone: 2C11]
Antistoffer
Type: Primary
Antigen: POU1F1 (POU class 1 homeobox 1)
Clonality: monoclonal
Clone: 2C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR