Anti-Rb2 p130 Mouse Monoclonal Antibody [clone: KAB40]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10576-50EA 4140 DKK
ABCAAB10576-50
Anti-Rb2 p130 Mouse Monoclonal Antibody [clone: KAB40]
Type: Primary
Antigen: Rb2 p130
Clonality: monoclonal
Clone: KAB40
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR