Anti-CDC25C Mouse Monoclonal Antibody [clone: DCS-193]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10537-50EA 4300 DKK
ABCAAB10537-50
Anti-CDC25C Mouse Monoclonal Antibody [clone: DCS-193]
Type: Primary
Antigen: CDC25C (cell division cycle 25C)
Clonality: monoclonal
Clone: DCS-193
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR