Anti-BIN1 Mouse Monoclonal Antibody [clone: 99D]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10532-50EA 4510 DKK
ABCAAB10532-50
Anti-BIN1 Mouse Monoclonal Antibody [clone: 99D]
Type: Primary
Antigen: BIN1 (bridging integrator 1)
Clonality: monoclonal
Clone: 99D
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR