Anti-TGF beta 1 Chicken Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10518-25EA 4650 DKK
ABCAAB10518-25
Anti-TGF beta 1 Chicken Polyclonal Antibody
Type: Primary
Antigen: TGF beta 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR