Anti-UBE3A Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB10488-50EA 4098.45 DKK
ABCAAB10488-50
Anti-UBE3A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: UBE3A (ubiquitin protein ligase E3A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human