Anti-CCL6 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10351-100EA 4140 DKK
ABCAAB10351-100
Anti-CCL6 Goat Polyclonal Antibody
Type: Primary
Antigen: CCL6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR