Anti-Endomorphin 2 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10289-50EA 4830 DKK
ABCAAB10289-50
Anti-Endomorphin 2 Rabbit Polyclonal Antibody
Type: Primary
Antigen: Endomorphin 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR