Anti-Fibrinogen Mouse Monoclonal Antibody HRP (Horseradish Peroxidase) [clone: 40F11]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10068-200EA 4720 DKK
ABCAAB10068-200
Anti-Fibrinogen Mouse Monoclonal Antibody HRP (Horseradish Peroxidase) [clone: 40F11]
Type: Primary
Antigen: Fibrinogen (fibrinogen)
Clonality: monoclonal
Clone: 40F11
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR