Anti-Cardiac FABP Mouse Monoclonal Antibody [clone: 6B6]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10059-250EA 0 DKK
ABCAAB10059-250
Anti-Cardiac FABP Mouse Monoclonal Antibody [clone: 6B6]
Type: Primary
Antigen: Cardiac FABP
Clonality: monoclonal
Clone: 6B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR