Anti-Creatine Kinase MB Mouse Monoclonal Antibody [clone: CK-10]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10045-100EA 4650 DKK
ABCAAB10045-100
Anti-Creatine Kinase MB Mouse Monoclonal Antibody [clone: CK-10]
Type: Primary
Antigen: Creatine Kinase MB
Clonality: monoclonal
Clone: CK-10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR