Anti-Corticotropin Releasing Factor Mouse Monoclonal Antibody [clone: 5D5]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10035-250EA 4650 DKK
ABCAAB10035-250
Anti-Corticotropin Releasing Factor Mouse Monoclonal Antibody [clone: 5D5]
Type: Primary
Antigen: Corticotropin Releasing Factor
Clonality: monoclonal
Clone: 5D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR