Anti-MUC16 Mouse Monoclonal Antibody [clone: X325]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB10033-500EA 5570 DKK
ABCAAB10033-500
Anti-MUC16 Mouse Monoclonal Antibody [clone: X325]
Antistoffer
Type: Primary
Antigen: MUC16 (mucin 16, cell surface associated)
Clonality: monoclonal
Clone: X325
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR