Anti-IL3 beta Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9874-100EA 4250 DKK
ABCAAB9874-100
Anti-IL3 beta Goat Polyclonal Antibody
Type: Primary
Antigen: IL3 beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR