Anti-IL3 Mouse Monoclonal Antibody [clone: 1F4]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9814-100EA 4250 DKK
ABCAAB9814-100
Anti-IL3 Mouse Monoclonal Antibody [clone: 1F4]
Type: Primary
Antigen: IL3 (interleukin 3 (colony-stimulating factor, multiple))
Clonality: monoclonal
Clone: 1F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR