Anti-SDF1 beta Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9799-100EA 4610 DKK
ABCAAB9799-100
Anti-SDF1 beta Goat Polyclonal Antibody
Type: Primary
Antigen: SDF1 beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR