Anti-NTF3 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9791-100EA 4300 DKK
ABCAAB9791-100
Anti-NTF3 Goat Polyclonal Antibody
Type: Primary
Antigen: NTF3 (neurotrophin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR