Anti-IL4 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9728-100EA 5330 DKK
ABCAAB9728-100
Anti-IL4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR